Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250991.1 | NP_001237920.1 | 7 - 8 | 1047 | 93 | |
GenBank mRNA | AJ006308.1 | - | 7 - 8 | 1047 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase Glycine max Probe spans exons |