Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249285.1 | NP_001236214.1 | 17 - 18 | 2291 | 87 | |
GenBank mRNA | AJ276866.1 | - | 18 - 19 | 2465 | 87 | |
AY230156.1 | - | 17 - 18 | 2291 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Glycine max Probe spans exons |