Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ401230.1 | - | 6 - 7 | 1161 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Glycine max Probe spans exons |