Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249369.1 | NP_001236298.1 | 3 - 4 | 1399 | 70 | |
GenBank mRNA | AJ563364.1 | - | 3 - 4 | 1399 | 70 | |
AK244954.1 | - | 3 - 4 | 1445 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter 3' Most Detect Genomic DNA Glycine max Probe spans exons |