Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY017473.1 | - | 5 - 6 | 1661 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Glycine max Probe spans exons |