Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249662.1 | NP_001236591.1 | 1 - 2 | 249 | 71 | |
GenBank mRNA | AK245291.1 | - | 1 - 2 | 242 | 71 | |
AY095999.1 | - | 1 - 2 | 249 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |