Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250327.1 | NP_001237256.1 | 7 - 8 | 1101 | 68 | |
GenBank mRNA | AY263347.1 | - | 7 - 8 | 1101 | 68 | |
BT097439.1 | - | 8 - 9 | 1201 | 68 | ||
FJ014778.1 | - | 6 - 7 | 966 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase 3' Most Detect Genomic DNA Glycine max Probe spans exons |