Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255528.1 | NP_001242457.1 | 6 - 7 | 792 | 85 | |
GenBank mRNA | AY652765.1 | - | 5 - 6 | 454 | 85 | |
HQ636610.1 | - | 6 - 7 | 792 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |