Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250756.1 | NP_001237685.1 | 3 - 4 | 826 | 64 | |
GenBank mRNA | D13949.1 | - | 3 - 4 | 826 | 64 | |
J03211.1 | - | 3 - 4 | 852 | 64 | ||
JQ929619.1 | - | 3 - 4 | 771 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Glycine max Probe spans exons |