Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251273.1 | NP_001238202.1 | 4 - 5 | 409 | 88 | |
GenBank mRNA | L28002.1 | - | 4 - 5 | 409 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |