Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249944.1 | NP_001236873.1 | 11 - 12 | 1058 | 124 | |
GenBank mRNA | X95582.1 | - | 11 - 12 | 1058 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Glycine max Probe spans exons |