Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250768.1 | NP_001237697.1 | 6 - 7 | 967 | 107 | |
GenBank mRNA | AM261476.1 | - | 6 - 7 | 967 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Glycine max Probe spans exons |