Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRY1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248276.1 | NP_001235205.1 | 3 - 4 | 1265 | 81 | |
GenBank mRNA | AB498928.1 | - | 3 - 4 | 1265 | 81 | |
DQ401046.1 | - | 3 - 4 | 1239 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Glycine max Probe spans exons |