Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249446.1 | NP_001236375.1 | 1 - 2 | 803 | 100 | |
GenBank mRNA | AJ875407.1 | - | 1 - 2 | 803 | 100 | |
AK245571.1 | - | 1 - 2 | 820 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |