Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251411.1 | NP_001238340.1 | 2 - 3 | 677 | 91 | |
GenBank mRNA | AM075829.1 | - | 2 - 3 | 677 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |