Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251411.1 | NP_001238340.1 | 8 - 9 | 1341 | 72 | |
GenBank mRNA | AM075829.1 | - | 8 - 9 | 1341 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |