Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251661.1 | NP_001238590.1 | 2 - 3 | 945 | 91 | |
GenBank mRNA | DQ136026.1 | - | 2 - 3 | 945 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Glycine max Probe spans exons |