Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK244720.1 | - | 2 - 3 | 265 | 63 | |
DQ822969.1 | - | 1 - 2 | 171 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Glycine max Probe spans exons |