Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254057.2 | NP_001240986.1 | 3 - 4 | 629 | 84 | |
GenBank mRNA | BT095976.1 | - | 3 - 4 | 631 | 84 | |
EU019561.1 | - | 2 - 3 | 242 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Glycine max Probe spans exons |