Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK245515.1 | - | 5 | 1542 | 80 | |
EU019591.1 | - | 3 | 746 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Glycine max Both primers and probe map within a single exon |