Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251615.1 | NP_001238544.1 | 3 | 849 | 77 | |
GenBank mRNA | FJ393222.1 | - | 3 | 849 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Glycine max Both primers and probe map within a single exon |