Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000507638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000507638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000507638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000507638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024330.hg.1 |
TC04001765.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024330.hg.1 |
ENST00000280193.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024330.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000507638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000507638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000507638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000507638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012487.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185624.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185624.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185624.hg.1 |
ENST00000618562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185624.hg.1 |
NM_005429.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185624.hg.1 |
TC0400012487.hg.1 |
Clariom D TC |