Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002600.hg.1 |
TC01000155.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002600.hg.1 |
ENST00000376259.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002600.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002600.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002601.hg.1 |
TC01000155.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002601.hg.1 |
ENST00000376259.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002601.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002601.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000489921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000489921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000489921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000489921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000584158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000622420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
ENST00000622420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NR_039775.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NR_107000.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006881.hg.1 |
NR_107000.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147940.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147940.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147940.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147940.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147940.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147940.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147940.hg.1 |
TC0100006881.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000489921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000489921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000489921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000489921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000536782.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000584158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000622420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
ENST00000622420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NR_039775.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NR_107000.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006881.hg.1 |
NR_107000.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147939.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147939.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147939.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147939.hg.1 |
ENST00000376259.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147939.hg.1 |
ENST00000492361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147939.hg.1 |
NM_001066.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147939.hg.1 |
TC0100006881.hg.1 |
Clariom D TC |