Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018175.hg.1 |
TC0X001357.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018175.hg.1 |
ENST00000308167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018175.hg.1 |
ENST00000335997.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018175.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018175.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018176.hg.1 |
TC0X001357.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018176.hg.1 |
ENST00000308167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018176.hg.1 |
ENST00000335997.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018176.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018176.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173249.hg.1 |
TC0X00010799.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173248.hg.1 |
TC0X00010799.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000308167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000335997.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000434609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
ENST00000615377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001127197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010799.hg.1 |
NM_001421.3 |
Refseq |