Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000526981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000526981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
ENST00000526981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047210.hg.1 |
TC01003029.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047210.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047210.hg.1 |
ENST00000369489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047210.hg.1 |
ENST00000457047.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047210.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047210.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047210.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215067.hg.1 |
TC0100015397.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369487.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000369489.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000457047.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000464088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000526981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000526981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
ENST00000526981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001144822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NM_001779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015397.hg.1 |
NR_026665.1 |
Refseq |