Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001009.hg.1 |
ENST00000262958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001009.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001009.hg.1 |
TC19000065.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001010.hg.1 |
ENST00000262958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001010.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001010.hg.1 |
TC19000065.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001007.hg.1 |
ENST00000262958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001007.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001007.hg.1 |
TC19000065.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001008.hg.1 |
ENST00000262958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001008.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001008.hg.1 |
TC19000065.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000586082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000586082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000586082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146241.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146241.hg.1 |
TC1900006620.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146241.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146243.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146243.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146243.hg.1 |
TC1900006620.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146243.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146243.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146247.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146247.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146247.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146247.hg.1 |
TC1900006620.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146247.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146247.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146247.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146245.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146245.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146245.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146245.hg.1 |
TC1900006620.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146245.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146245.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146245.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146242.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146242.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146242.hg.1 |
TC1900006620.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146242.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146242.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000262958.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000586082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000586082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000586082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
ENST00000592455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006620.hg.1 |
NM_002068.3 |
Refseq |