Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030185.hg.1 |
TC03001673.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030185.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030186.hg.1 |
TC03001673.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030186.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030187.hg.1 |
TC03001673.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030187.hg.1 |
ENST00000490035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030187.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030190.hg.1 |
TC03001673.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030190.hg.1 |
ENST00000490035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030190.hg.1 |
ENST00000539563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030190.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030195.hg.1 |
TC03001673.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030195.hg.1 |
ENST00000490035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030195.hg.1 |
ENST00000539563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030195.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189937.hg.1 |
ENST00000490035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189937.hg.1 |
ENST00000539563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189937.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189937.hg.1 |
ENST00000490035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189937.hg.1 |
ENST00000539563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189937.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189937.hg.1 |
TC0300012073.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300189941.hg.1 |
ENST00000333617.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189941.hg.1 |
ENST00000475403.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189941.hg.1 |
ENST00000490035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189941.hg.1 |
ENST00000539563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189941.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189941.hg.1 |
ENST00000333617.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189941.hg.1 |
ENST00000475403.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189941.hg.1 |
ENST00000490035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189941.hg.1 |
ENST00000539563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189941.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189941.hg.1 |
TC0300012073.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
ENST00000333617.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
ENST00000473171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
ENST00000475403.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
ENST00000490035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
ENST00000539563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
ENST00000333617.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
ENST00000473171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
ENST00000475403.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
ENST00000490035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
ENST00000539563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189946.hg.1 |
TC0300012073.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300189938.hg.1 |
ENST00000333617.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189938.hg.1 |
ENST00000490035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189938.hg.1 |
ENST00000539563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189938.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189938.hg.1 |
ENST00000333617.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189938.hg.1 |
ENST00000490035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189938.hg.1 |
ENST00000539563.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189938.hg.1 |
NM_002338.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189938.hg.1 |
TC0300012073.hg.1 |
Clariom D TC |