Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000495636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000495636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000495636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000495636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
ENST00000495636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001380.hg.1 |
TC03000063.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001380.hg.1 |
ENST00000287766.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001380.hg.1 |
ENST00000536032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001380.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000425938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000462473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000495636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000495636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000495636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000495636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
ENST00000495636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006599.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145801.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145801.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145801.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145801.hg.1 |
ENST00000287766.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145801.hg.1 |
ENST00000460480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145801.hg.1 |
NM_003042.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145801.hg.1 |
TC0300006599.hg.1 |
Clariom D TC |