Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011580.hg.1 |
TC06000983.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011580.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011580.hg.1 |
ENST00000368087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011580.hg.1 |
ENST00000476845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011580.hg.1 |
ENST00000489091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011580.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011580.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011583.hg.1 |
TC06000983.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011583.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011583.hg.1 |
ENST00000368087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011583.hg.1 |
ENST00000476845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011583.hg.1 |
ENST00000489091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011583.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011583.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011581.hg.1 |
TC06000983.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011581.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011581.hg.1 |
ENST00000368087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011581.hg.1 |
ENST00000476845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011581.hg.1 |
ENST00000489091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011581.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011581.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000484820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000484820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000484820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000484820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164829.hg.1 |
TC0600009455.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000484820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000484820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000484820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000484820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009455.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
ENST00000275196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
ENST00000469293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164831.hg.1 |
TC0600009455.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
ENST00000484820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
ENST00000356962.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
ENST00000368087.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
ENST00000484820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
ENST00000498260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
NM_000045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
NM_001244438.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164833.hg.1 |
TC0600009455.hg.1 |
Clariom D TC |