Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057211.hg.1 |
TC01003726.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057211.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057211.hg.1 |
ENST00000367195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057211.hg.1 |
ENST00000545733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057211.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057212.hg.1 |
TC01003726.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057212.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057212.hg.1 |
ENST00000367195.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057212.hg.1 |
ENST00000545733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057212.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228934.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228934.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228934.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228934.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228934.hg.1 |
TC0100017020.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100228932.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228932.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228932.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228932.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228932.hg.1 |
TC0100017020.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100228933.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228933.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228933.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228933.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228933.hg.1 |
TC0100017020.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
ENST00000272190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017020.hg.1 |
NM_000537.3 |
Refseq |