Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013038.hg.1 |
TC0X000978.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013038.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013038.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013038.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013041.hg.1 |
TC0X000978.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013041.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013037.hg.1 |
TC0X000978.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013037.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013037.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013037.hg.1 |
ENST00000536181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013037.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013030.hg.1 |
TC0X000978.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013030.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013030.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013030.hg.1 |
ENST00000536181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013030.hg.1 |
ENST00000538942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013030.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013036.hg.1 |
TC0X000978.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013036.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013036.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013036.hg.1 |
ENST00000536181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013036.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164816.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164816.hg.1 |
ENST00000468431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164816.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164816.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164816.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164816.hg.1 |
ENST00000468431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164816.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164816.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164816.hg.1 |
TC0X00009486.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00164818.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164818.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164818.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164818.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164818.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164818.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164818.hg.1 |
TC0X00009486.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164821.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164821.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164821.hg.1 |
TC0X00009486.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00164812.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164812.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164812.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164812.hg.1 |
ENST00000378069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164812.hg.1 |
ENST00000487544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164812.hg.1 |
NM_000898.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164812.hg.1 |
TC0X00009486.hg.1 |
Clariom D TC |