Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007505.hg.1 |
TC0X000570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007505.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007505.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007506.hg.1 |
TC0X000570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007506.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007506.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008207.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155996.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155996.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155996.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155996.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155996.hg.1 |
TC0X00008207.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00155998.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155998.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155998.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155998.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155998.hg.1 |
TC0X00008207.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00155997.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155997.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155997.hg.1 |
ENST00000371906.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155997.hg.1 |
NM_000686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155997.hg.1 |
TC0X00008207.hg.1 |
Clariom D TC |