Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001339.hg.1 |
ENST00000300177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001339.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001339.hg.1 |
TC15000226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001342.hg.1 |
ENST00000300177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001342.hg.1 |
ENST00000322805.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001342.hg.1 |
ENST00000560677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001342.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001342.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001342.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001342.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001342.hg.1 |
TC15000226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001333.hg.1 |
ENST00000300177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001333.hg.1 |
ENST00000560677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001333.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001333.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001333.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001333.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001333.hg.1 |
TC15000226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001341.hg.1 |
ENST00000300177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001341.hg.1 |
ENST00000322805.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001341.hg.1 |
ENST00000560677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001341.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001341.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001341.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001341.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001341.hg.1 |
TC15000226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001344.hg.1 |
ENST00000300177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001344.hg.1 |
ENST00000322805.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001344.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001344.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001344.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001344.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001344.hg.1 |
TC15000226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001337.hg.1 |
ENST00000300177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001337.hg.1 |
ENST00000322805.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001337.hg.1 |
ENST00000560677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001337.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001337.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001337.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001337.hg.1 |
TC15000226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001336.hg.1 |
ENST00000300177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001336.hg.1 |
ENST00000322805.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001336.hg.1 |
ENST00000560677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001336.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001336.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001336.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001336.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001336.hg.1 |
TC15000226.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006788.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147181.hg.1 |
TC1500006788.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147173.hg.1 |
TC1500006788.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147182.hg.1 |
TC1500006788.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500147179.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147179.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147179.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147179.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147179.hg.1 |
TC1500006788.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147176.hg.1 |
TC1500006788.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500147177.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147177.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147177.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147177.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147177.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147177.hg.1 |
ENST00000560677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147177.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147177.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147177.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147177.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147177.hg.1 |
TC1500006788.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500147184.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147184.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147184.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147184.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147184.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147184.hg.1 |
ENST00000560830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147184.hg.1 |
ENST00000622074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147184.hg.1 |
NM_001191322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147184.hg.1 |
NM_001191323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147184.hg.1 |
NM_013372.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147184.hg.1 |
TC1500006788.hg.1 |
Clariom D TC |