Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000261332.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000261332.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000261332.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000261332.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000261332.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000261332.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000261332.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000399061.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000399061.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000399061.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000399061.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000399061.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000399061.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000399061.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000589539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000589539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000589539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000589881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000589881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000589881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000589881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000589881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000589881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000589881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000590140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000590140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000590140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000590140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000590140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000593130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000593130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
ENST00000593130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_001308123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_001308123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_001308123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_001308123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_001308123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_001308123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_006965.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_006965.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_006965.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_006965.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_006965.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_006965.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008458.hg.1 |
NM_006965.3 |
Refseq |