Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041114.hg.1 |
TC01002627.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041114.hg.1 |
ENST00000294338.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041114.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041114.hg.1 |
ENST00000491793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01041114.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000491793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000491793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000491793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
ENST00000491793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205409.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205409.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205409.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205409.hg.1 |
ENST00000491793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205409.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205409.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205409.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205409.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205409.hg.1 |
ENST00000491793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205409.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205409.hg.1 |
TC0100014081.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000294338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000371885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000489919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000491793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000491793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000491793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
ENST00000491793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014081.hg.1 |
NM_005764.3 |
Refseq |