Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000528540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000528540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013302.hg.1 |
ENST00000392883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013302.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013302.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013302.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013302.hg.1 |
TC11001061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013298.hg.1 |
ENST00000392883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013298.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013298.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013298.hg.1 |
TC11001061.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000528540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000528540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100168392.hg.1 |
TC1100009202.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000292144.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000392883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000527777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000528540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000528540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000532917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
ENST00000533462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009202.hg.1 |
NM_000073.2 |
Refseq |