Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015427.hg.1 |
TC01000968.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015427.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015427.hg.1 |
ENST00000429072.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015427.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015427.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015427.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000464329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000464329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000464329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000464329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000471220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000471220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000471220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000471220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167546.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167546.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167546.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167546.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167546.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167546.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167546.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167546.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167546.hg.1 |
TC0100009394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000271324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000464329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000464329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000464329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000464329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000471220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000471220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000471220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000471220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000476408.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
ENST00000497404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_000560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009394.hg.1 |
NM_001040033.1 |
Refseq |