Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025463.hg.1 |
ENST00000264563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025463.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025463.hg.1 |
TC19001870.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011467.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
NM_001267718.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
TC1900011467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
ENST00000264563.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
ENST00000585513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
ENST00000587093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
ENST00000590625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
NM_000641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197208.hg.1 |
NM_001267718.1 |
Refseq |