Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017990.hg.1 |
ENST00000282091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017990.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017990.hg.1 |
TC11001435.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017989.hg.1 |
ENST00000282091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017989.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017989.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017989.hg.1 |
TC11001435.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010157.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176995.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176995.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176995.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176995.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176995.hg.1 |
TC1100010157.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100176993.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176993.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176993.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176993.hg.1 |
ENST00000282091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176993.hg.1 |
ENST00000529816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176993.hg.1 |
NM_000315.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176993.hg.1 |
TC1100010157.hg.1 |
Clariom D TC |