Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018132.hg.1 |
TC0X001354.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018132.hg.1 |
ENST00000307484.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018132.hg.1 |
ENST00000429967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018132.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010787.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010787.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010787.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010787.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010787.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010787.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010787.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010787.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018131.hg.1 |
TC0X001354.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018131.hg.1 |
ENST00000307484.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018131.hg.1 |
ENST00000429967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018131.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018130.hg.1 |
TC0X001354.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018130.hg.1 |
ENST00000307484.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018130.hg.1 |
ENST00000427399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018130.hg.1 |
ENST00000429967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018130.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173168.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173168.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173168.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173168.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173168.hg.1 |
TC0X00010787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010787.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010787.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010787.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010787.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010787.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010787.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010787.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010787.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173169.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173169.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173169.hg.1 |
ENST00000429967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173169.hg.1 |
NM_017413.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173169.hg.1 |
TC0X00010787.hg.1 |
Clariom D TC |