Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020679.hg.1 |
ENST00000253054.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020679.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19020679.hg.1 |
TC19001517.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000594700.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000594700.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000594700.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000594700.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000594700.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000598218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
TC1900010651.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900187867.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000253054.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000594700.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000594700.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000594700.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000594700.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000594700.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000595636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000596583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000597595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000598218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000600322.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000601731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
ENST00000602185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_001301008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010651.hg.1 |
NM_004877.3 |
Refseq |