Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016939.hg.1 |
TC0X001256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016939.hg.1 |
ENST00000327674.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016939.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016939.hg.1 |
ENST00000487487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016939.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171179.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171179.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171179.hg.1 |
ENST00000487487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171179.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171179.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171179.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171179.hg.1 |
ENST00000487487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171179.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171179.hg.1 |
TC0X00010452.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |