Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |