Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000549790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000549790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000549790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000549790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000553019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000553019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000553019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
ENST00000553019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023936.hg.1 |
ENST00000322349.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023936.hg.1 |
ENST00000540777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023936.hg.1 |
ENST00000547833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023936.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023936.hg.1 |
TC12001811.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200189315.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189315.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189315.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189315.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189315.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189315.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189315.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189315.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189315.hg.1 |
TC1200011519.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000322349.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000418984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000547833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000549790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000549790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000549790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000549790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000553019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000553019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000553019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
ENST00000553019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011519.hg.1 |
NM_003566.3 |
Refseq |