Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000365.hg.1 |
TC03000015.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000365.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000365.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000460806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000460806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000460806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000460806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000460806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000460806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000460806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000460806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006483.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144529.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144529.hg.1 |
ENST00000460806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144529.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144529.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144529.hg.1 |
ENST00000256495.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144529.hg.1 |
ENST00000460806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144529.hg.1 |
ENST00000467610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144529.hg.1 |
NM_003670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144529.hg.1 |
TC0300006483.hg.1 |
Clariom D TC |