Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001174.hg.1 |
ENST00000248121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001174.hg.1 |
ENST00000320633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001174.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001174.hg.1 |
TC16000056.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001175.hg.1 |
ENST00000248121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001175.hg.1 |
ENST00000320633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001175.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001175.hg.1 |
TC16000056.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001176.hg.1 |
ENST00000248121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001176.hg.1 |
ENST00000320633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001176.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001176.hg.1 |
TC16000056.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001173.hg.1 |
ENST00000248121.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001173.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001173.hg.1 |
TC16000056.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000564642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006580.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
ENST00000562045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
ENST00000563869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146107.hg.1 |
TC1600006580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600146100.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146100.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146100.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146100.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146100.hg.1 |
TC1600006580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600146104.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146104.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146104.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146104.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146104.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146104.hg.1 |
ENST00000568896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146104.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146104.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146104.hg.1 |
TC1600006580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600146102.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146102.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146102.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146102.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146102.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146102.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146102.hg.1 |
TC1600006580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600146101.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146101.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146101.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146101.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146101.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146101.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146101.hg.1 |
TC1600006580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600146103.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146103.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146103.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146103.hg.1 |
ENST00000248121.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146103.hg.1 |
ENST00000618464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146103.hg.1 |
NM_004209.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146103.hg.1 |
TC1600006580.hg.1 |
Clariom D TC |