Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023135.hg.1 |
TC02004920.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023135.hg.1 |
ENST00000244815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023135.hg.1 |
ENST00000289175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023135.hg.1 |
ENST00000308482.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023135.hg.1 |
ENST00000391999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023135.hg.1 |
ENST00000392000.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023135.hg.1 |
NM_001137550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023135.hg.1 |
NM_001137551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023135.hg.1 |
NM_001137552.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023135.hg.1 |
NM_001137553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023135.hg.1 |
NM_004735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000244815.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000289175.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000308482.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000392000.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000420665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000498053.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
NM_001137550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
NM_001137551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
NM_001137552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
NM_001137553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
NM_004735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000244815.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000289175.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000308482.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000392000.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000420665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
ENST00000498053.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
NM_001137550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
NM_001137551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
NM_001137552.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
NM_001137553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
NM_004735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225139.hg.1 |
TC0200016603.hg.1 |
Clariom D TC |