Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000601101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000601101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000601101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000601101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000601101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000602059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000602059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
ENST00000602059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000601101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000601101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000601101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000601101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000601101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000602059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000602059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
ENST00000602059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011857.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203884.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203884.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203884.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203884.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203884.hg.1 |
NM_005001.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203884.hg.1 |
TC1900011857.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203884.hg.1 |
ENST00000301457.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203884.hg.1 |
ENST00000593729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203884.hg.1 |
ENST00000595856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203884.hg.1 |
ENST00000601258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203884.hg.1 |
NM_005001.4 |
Refseq |