Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001722.hg.1 |
ENST00000343118.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001722.hg.1 |
ENST00000380618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001722.hg.1 |
ENST00000380620.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001722.hg.1 |
ENST00000398714.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001722.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001722.hg.1 |
NM_033170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001722.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001722.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001722.hg.1 |
TC21000177.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008520.hg.1 |
ENST00000475838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008520.hg.1 |
ENST00000475838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000343118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000343118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000343118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000343118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000343118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000398714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000398714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000398714.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158270.hg.1 |
TC2100008521.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100158270.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158270.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158270.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158270.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008520.hg.1 |
ENST00000475838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008520.hg.1 |
ENST00000475838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000343118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000343118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000343118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000343118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000343118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000380618.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000398714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000398714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000398714.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
ENST00000615480.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_001278650.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_006057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008521.hg.1 |
NM_033172.2 |
Refseq |