Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011679.hg.1 |
ENST00000228928.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011679.hg.1 |
ENST00000323881.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011679.hg.1 |
ENST00000546638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011679.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011679.hg.1 |
TC12000885.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000546638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000546638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000546638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000546800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000546800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000546800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000546973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000546973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000546973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000546973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000548514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000548514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000548514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000548514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000548514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000549918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000549918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000549918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000549918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000551007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000551007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000551007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
ENST00000551007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000546638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000546638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000546638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000546800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000546800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000546800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000546973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000546973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000546973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000546973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000548514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000548514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000548514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000548514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000548514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000549918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000549918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000549918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000549918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000551007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000551007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000551007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
ENST00000551007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008920.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166291.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166291.hg.1 |
ENST00000546638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166291.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166291.hg.1 |
ENST00000228928.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166291.hg.1 |
ENST00000546638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166291.hg.1 |
NM_006187.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166291.hg.1 |
TC1200008920.hg.1 |
Clariom D TC |